Learn More
Tenak™ ECO AluRack for Upright Freezers, 75 mm Box Height
ECO AluRack fits into any upright freezer and can be used with all standard cryoboxes.
78.95€ - 107.00€
Specifications
Description | Seawater resistant |
---|---|
Material | Aluminum |
For Use With (Equipment) | Upright Freezers |
Height (Metric) | 75 mm (Box) |
Product Code | Brand | Holds | Array | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Array | Price | Quantity & Availability | ||||
12402530
|
Tenak™
TE11260 |
Boxes (6) | 3 x 2 |
78.95€
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
||||
12412530
|
Tenak™
TE11262 |
Boxes (6) | 2 x 3 |
81.20€
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
12412540
|
Tenak™
TE11276 |
Boxes (16) | 4 x 4 |
92.00€
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
||||
12432530
|
Tenak™
TE11264 |
Boxes (9) | 3 x 3 |
83.50€
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
||||
12432540
|
Tenak™
TE11278 |
Boxes (20) | 5 x 4 |
103.00€
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
||||
12452530
|
Tenak™
TE11266 |
Boxes (12) | 4 x 3 |
88.60€
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
||||
12452540
|
Tenak™
TE11284 |
Boxes (10) | 2 x 5 |
92.00€
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
12472530
|
Tenak™
TE11272 |
Boxes (8) | 2 x 4 |
83.50€
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
12492530
|
Tenak™
TE11274 |
Boxes (12) | 3 x 4 |
82.35€
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
||||
12492540
|
Tenak™
TE11288 |
Boxes (20) | 4 x 5 |
107.00€
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
||||
Description
- Made of seawater resistant aluminum
- Fits into any freezer and can be used with all standard cryoboxes
- Racks can be used with all freezer brands
- Without white cardboard boxes
- Holds is expressed by Height x Dept
Specifications
Seawater resistant | |
Aluminum | |
Upright Freezers | |
75 mm (Box) |