Learn More
Tenak™ ECO AluRack for Upright Freezers, 32 mm Box Height, with Boxes
ECO AluRack fits into any upright freezer and can be used with all standard cryoboxes.
Brand:  Tenak™ TE11124B
Additional Details : Weight : 3.30000kg
 View more versions of this product
Product Code. 12422480
  • 156.00€ / Each
Estimated Shipment: 23-09-2024
Description
  • Made of seawater resistant aluminum
  • Fits into any freezer and can be used with all standard cryoboxes
  • Racks can be used with all freezer brands
  • Includes standard white cardboard boxes with 9 x 9 cell dividers
  • Holds is expressed by Height x Dept
Specifications
Specifications
White cardboard boxes, 9 x 9 cell dividers
7 x 3
Aluminum
425 x 139 x 252 mm
Seawater resistant
Boxes (21)
Upright Freezers
32 mm (Box)