Learn More
Tenak™ Eco AluRack for Upright Freezer, for Microtiterplates, 16 mm Shelf Height
Eco AluRack for upright freezers is designed to hold microtiterplates.
Brand:  Tenak™ TE15456
Additional Details : Weight : 1.60000kg
 View more versions of this product
Product Code. 12498220
  • 200.00€ / Each
Estimated Shipment: 26-08-2024
Description
  • For use in upright freezers
  • Racks can either be used as one plate per shelf or the plates can be stacked
  • Constructed of aluminum
Specifications
Specifications
19 x 5
Aluminum
450 x 135 x 323 mm
Microtiterplates (95)
Upright Freezers
16 mm