Learn More
Tenak™ Eco AluRack for Chest Freezer, for Microtiterplates, 16 mm Shelf Height
Eco AluRack for chest freezers is designed to hold microtiterplates.
90.85€ - 158.00€
Specifications
Material | Aluminum |
---|---|
For Use With (Equipment) | Chest Frseezers |
Height (Metric) | 16 mm |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
12408170
|
Tenak™
TE15108 |
Microtiterplates (29) |
129.00€
Each |
Estimated Shipment: 11-09-2024
Log in to see stock. |
|||||
12418170
|
Tenak™
TE15110 |
Microtiterplates (32) |
138.00€
Each |
Estimated Shipment: 11-09-2024
Log in to see stock. |
|||||
12428170
|
Tenak™
TE15112 |
Microtiterplates (35) |
148.00€
Each |
Estimated Shipment: 11-09-2024
Log in to see stock. |
|||||
12438170
|
Tenak™
TE15114 |
Microtiterplates (39) |
158.00€
Each |
Estimated Shipment: 11-09-2024
Log in to see stock. |
|||||
12468160
|
Tenak™
TE15100 |
Microtiterplates (16) |
90.85€
Each |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
12478160
|
Tenak™
TE15102 |
Microtiterplates (19) |
99.95€
Each |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
12488160
|
Tenak™
TE15104 |
Microtiterplates (22) |
110.00€
Each |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
12498160
|
Tenak™
TE15106 |
Microtiterplates (26) |
120.00€
Each |
Estimated Shipment: 11-09-2024
Log in to see stock. |
|||||
Description
- For use in chest freezers
- Racks can either be used as one plate per shelf or the plates can be stacked
- Constructed of aluminum
Specifications
Aluminum | |
Chest Frseezers | |
16 mm |