Learn More
Thermo Scientific™ EasyStart™ PCR Mix-in-a-Tube
Reduce tedious PCR set-up protocols to a single step using Thermo Scientific™ EasyStart™ PCR Mix-in-a-tube.
Specifications
| Volume (Metric) | 0.2 mL |
|---|---|
| Storage Requirements | Tubes can be stored at ambient temperatures for extended periods; To remove PCR product, simply pierce the wax layer with a pipet tip |
Description
Reduce tedious PCR set-up protocols to a single step using Thermo Scientific™ EasyStart™ PCR Mix-in-a-tube.
Features:
- Delivers better specificity and yield than conventionally prepared mixtures
- Compatible with a wide range of enzymes
- Wax layer provides safe, easy storage of reagent mixture, prevents oxidation and evaporation, and eliminates wasted reagents
Ordering Information:
Use 0.2mL tubes for 0.2mL heating blocks and 0.5mL tubes for 0.5 or 0.6mL heating blocks.
Specifications
| 0.2 mL | |
| Tubes can be stored at ambient temperatures for extended periods; To remove PCR product, simply pierce the wax layer with a pipet tip |