Learn More
Abeba™ Easy 7623 Shoes
Includes TPU Sole with high slip resistance. Upper leather and microfiber construction.
Brand:  Abeba™ 7623/35
Additional Details : Weight : 0.97000kg
 View more versions of this product
Product Code. 15612898
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Description
Description
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Smooth leather white/red
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3573)
  • Sanitized treated
  • Antistatic
  • Heel strap non-foldable, adjustable
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC
Leather
Slip-resistant
7623
White
35
Polyurethane