Learn More
Abeba™ Dynamic 7363 Shoes
Special respiration absorbing lining allows the foot to breathe. Includes TPU sole with high slip resistance.
Brand: Abeba™ 7363/37
Additional Details : Weight : 0.98000kg
Description
- CE EN ISO 20347:2012 OB A, E, FO SRC
- Microfiber
- Gray/Red
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3574)
- Sanitized treated
- Antistatic
- Heel strap non-foldable, adjustable
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC | |
Microfiber | |
Slip-resistant | |
7363 |
Gray, Red | |
37 | |
Polyurethane |