Learn More
Abeba™ Dynamic 7341 Shoes
Special respiration absorbing lining allows the foot to breathe. Includes TPU sole with high slip resistance.
Brand:  Abeba™ 7341/38
Additional Details : Weight : 0.99000kg
 View more versions of this product
Product Code. 15622738
  • 71.53€ / 1 pair
Estimated Shipment: 31-05-2024
Description
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Microfiber with breathable textile inlays
  • Black/Blue
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3574)
  • Sanitized treated
  • Antistatic
  • Heel strap non-foldable, adjustable
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC
Microfiber
Slip-resistant
7341
Black, Blue
38
Polyurethane