Learn More
Abeba™ Dynamic 7312 Shoes
Special respiration absorbing lining allows the foot to breathe. Includes TPU sole with high slip resistance.
Brand:  Abeba™ 7312/46
57.71 EUR valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Additional Details : Weight : 0.94000kg
 View more versions of this product
Product Code. 15662658
  • 63.95€ / 1 pair
Estimated Shipment: 03-06-2024
Description
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Smooth leather white/blue
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3574)
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC
Leather
Slip-resistant
7312
White
46
Polyurethane