Learn More
DWK Life Sciences DURAN™ Single way Stopcock, complete with hollow key with glass handle
To be used for higher tightness requirements.
Brand: DWK Life Sciences 286458804
Additional Details : Weight : 0.05000kg
Description
DURAN™ single way stopcocks complete with hollow DURAN™ cock plug are well suited for work with temperature changes and higher tightness requirements.
- Ground shut-off valve sleeve with hollow glass plug with glass handle
- High tightness
- Taper 1:10
- Glass articles for further processing
Specifications
2.5 mm | |
Borosilicate Glass (3.3) | |
14.5 NS | |
50/Pk. |
9 mm | |
Yes | |
1.5 mm | |
When working with temperature changes and higher tightness requirements; Installation in glass apparatus and devices |