Learn More
DWK Life Sciences DURAN™ Single way Stopcock, complete with DURAN™ key with groove
Ground shut-off taps without tap plug.On-and-off taps with DURAN™ grooved tap plugs.
169.00€ - 299.00€
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
Autoclavable | Yes |
For Use With (Application) | Installation in glass apparatus and devices |
Product Code | Brand | Closure Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Size | Price | Quantity & Availability | |||||
15951591
|
DWK Life Sciences
286407606 |
24 NS |
246.00€
Pack of 5 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||||
15864698
|
DWK Life Sciences
286402402 |
10 NS |
177.00€
Pack of 10 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||||
15844708
|
DWK Life Sciences
286407803 |
29.2 NS |
299.00€
Pack of 5 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||||
15834708
|
DWK Life Sciences
286407409 |
21.5 NS |
169.00€
Pack of 5 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||||
Description
DURAN™ single way stopcocks complete with DURAN™ grooved tap plugs are shut-off taps in which the DURAN™ grooved tap plug is secured via the O-ring.
- Ground shut-off valve sleeve complete with DURAN™ grooved plug
- Securing of the tap plug by means of O-ring
- Taper 1:10
- Glass articles for further processing
Specifications
Borosilicate Glass (3.3) | |
Yes | |
Installation in glass apparatus and devices |