Learn More
DWK Life Sciences DURAN™ Measuring Cylinder with Hexagonal Base, Class A, Blue Scale, Ring Graduations, with Batch Certificate and Certificate of Conformity
Product Code. 15280846 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Pack of 2
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15280846 100 mL Pack of 2
15270846 50 mL Pack of 2
15290846 250 mL Pack of 2
15200856 500 mL Pack of 2
15210856 1000 mL Each
15220856 2000 mL Each
15240846 5 mL Pack of 2
15250846 10 mL Pack of 2
15260846 25 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 15280846 Supplier DWK Life Sciences Supplier No. 213902402
  • ONLINE EXCLUSIVE PRICE
    28.77€
    25.19€ / Pack of 2
    Save 3.58€
    12% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
DURAN™ Measuring cylinders are for holding measuring varying amounts of liquids.
DURAN™ Measuring cylinders are for holding measuring varying amounts of liquids. Available in two different accuracy Classes – Class A with main ring points graduation and Class B with line graduation. For additional differentiation, the measuring cylinders with accuracy Class A have a blue graduation whereas the B Class measuring cylinders a white graduation. Volume content tolerances for measuring and mixing cylinders conform to DIN and ISO accuracy limits. Thus the desired concentration can be precisely created. Measuring cylinders with a batch number and accuracy Class A are supplied with a batch certificate. This certificate documents the mean value obtained from measuring the batch in question, the standard deviation and the day of issue. The batch certificates can also be retrieved online. The batch number consists of four digits. The first two numbers specify the production year, and the following two numbers specify the batch, e.g.: 18.01.
  • Calibration is based on the poured in volume (In) at a + 20°C reference temperature
  • Large hexagonal base equipped with three knobs which increase its stability and prevent the cylinder from rolling
  • A uniform wall thickness over the entire measuring range prevents wedge errors
  • Fired-on, highly durable blue print
  • Easy-to-read scale with graduation marks to estimate the content
  • With circular ring graduations
  • The volume content tolerances conform to accuracy Class A
Specifications
Material Borosilicate Glass (3.3)
Capacity (Metric) 100 mL
For Use With (Application) Measuring liquid amounts; Diluting solutions; Mixing several components; Volumetric measurement; To ensure a long service life for your volumetric glassware and to exclude possible volume changes, these glass products should not be heated above + 250°C in drying cabinets or sterilisers
Includes 2/Pk.
Scale Blue Graduated
Graduations 1 mL
Autoclavable Autoclavable
Sterility Non-sterile
Calibration To Contain
Base Type Hexagonal
Scale Color Blue
Class Class A
Height (Metric) 256 mm
Certifications/Compliance ISO 4788
Tolerance ±0.5 mL
Product Type Graduated Measuring Cylinder
Diameter (Metric) Outer 29 mm