missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Glass Stopper, Clear, Standard Taper Ground Joint, Octagonal flat head

Clear DURAN™ glass ground stopper.

Brand:  DWK Life Sciences 216240307

93.29 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.



Additional Details : Weight : 2.50000kg

 View more versions of this product

Product Code. 15814508

  • 98.20€ / Pack of 10
Estimated Shipment: 16-07-2024
to see stock.
Add to basket

Description

Description

Semi-hollow or solid DURAN™ glass stoppers can be used to close glassware items such as flasks, separatory funnels or reagent bottles with ground joint necks. The Standard Taper ground glass joint forms a tight, leak-proof seal. Use grease for heating or vacuum applications. With octagonal shaped head. Note on using DURAN™ glass stoppers with soda-lime glass bottles: If the bottle and the stopper have a temperature difference greater than 30°C, the glass stoppers can become stuck!

  • All DURAN™ glass components and therefore suitable for storage of aggressive materials
  • Tight closure due to Standard Taper ground joint
  • Octagonal shaped head
Specifications

Specifications

Storage of air sensitive reagents, and chemicals; Storage of hygroscopic solids, granules and powders; Storage of pharmaceutical APIs; Sealing laboratory glassware such as flasks, reagent bottles, and mixing cylinders
Boro 3.3
No
10/Pk.
ST 10/19
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers