Learn More
Witeg™ Duran Glass Separatory Funnels
Witeg™ Duran™ Glass Separatory Funnels are used in liquid-liquid extractions to separate (partition) the components of a mixture into two immiscible solvent phases of different densities.
Brand:  Witeg™ 2 677 010
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11873861
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Description
Description
  • Conical shape
  • Graduated
  • Economy-needle valve stopcock
  • PTFE-needle valve
  • Screw-thread and cap
Specifications
Specifications
100 mL
Duran Glass, Borosilicate Glass
1Pack
Separatory funnel