Learn More
Lenz Laborglasinstrumente™ Duran Glass Separating Funnels
Manufactured from borosilicate glass. Lenz Laborglasinstrumente™ Duran Glass Separating Funnels are used in liquid-liquid extractions to separate the components of a mixture into immiscible solvent phases of different densities.
Brand: Lenz Laborglasinstrumente™ B0462951
Additional Details : Weight : 0.08100kg
Description
- DURAN™ Borosilicate Glass
- For use with liquid extractions
Specifications
50 mL | |
Duran Glass, Borosilicate Glass |