Learn More
Lenz Laborglasinstrumente™ Duran Glass Separating Funnels
Manufactured from borosilicate glass. Lenz Laborglasinstrumente™ Duran Glass Separating Funnels are used in liquid-liquid extractions to separate the components of a mixture into immiscible solvent phases of different densities.
Brand:  Lenz Laborglasinstrumente™ B0462951
Additional Details : Weight : 0.08100kg
Product Code. 10274120
  • 63.00€ / Each
Estimated Shipment: 28-06-2024
Description
  • DURAN™ Borosilicate Glass
  • For use with liquid extractions
Specifications
Specifications
50 mL
Duran Glass, Borosilicate Glass