Learn More
Lenz Laborglasinstrumente™ Duran Glass Separating Funnels
Manufactured from borosilicate glass. Lenz Laborglasinstrumente™ Duran Glass Separating Funnels are used in liquid-liquid extractions to separate the components of a mixture into immiscible solvent phases of different densities.
Brand:  Lenz Laborglasinstrumente™ B0463001
Additional Details : Weight : 0.11800kg
Product Code. 10326210
  • 69.00€ / Each
Estimated Shipment: 21-06-2024
to see stock.
Add to basket
Description
Description
  • DURAN™ Borosilicate Glass
  • For use with liquid extractions
Specifications
Specifications
250 mL
Duran Glass, Borosilicate Glass