Learn More
Lenz Laborglasinstrumente™ Reduction Adapters with Ground Joints
Provides for secure and easy connection from a large cone to a small socket in glass tubing. Lenz Laborglasinstrumente™ Reduction Adapters with Ground Joints are manufactured from durable DURAN™ borosilicate glass.
Brand: Lenz Laborglasinstrumente™ 01502945
Additional Details : Weight : 0.05310kg
Description
- Acc. to DIN 12257
- High taper accuracy of the cones
- Minimized need for grease
- Excellent tightness
- Corrosion-free and resistant to thermal shock
Specifications
DIN 12257 and in addition to DIN standard | |
Duran Glass | |
1Each |
NS 45/40 | |
Reduction Adapter |