Learn More
Lenz Laborglasinstrumente™ Duran Glass ISO Form Separating Funnels
Created from borosilicate glass. Lenz Laborglasinstrumente™ Duran Glass ISO Form Separating Funnels are used in liquid-liquid extractions to separate the components of a mixture into immiscible solvent phases of different densities.
Brand:  Lenz Laborglasinstrumente™ B0463151
Additional Details : Weight : 0.40200kg
Product Code. 10723631
  • 128.00€ / Each
Estimated Shipment: 28-06-2024
Description
  • According to ISO
  • Material: DURAN™ Borosilicate Glass
  • For use with liquid extractions
Specifications
Specifications
1000 mL
Duran Glass, Borosilicate Glass