Learn More
Lenz Laborglasinstrumente™ Duran Glass ISO Form Separating Funnels
Lenz Laborglasinstrumente™ Duran Glass ISO Form Separating Funnels are used in liquid-liquid extractions to separate the components of a mixture into immiscible solvent phases of different densities.
Brand:  Lenz Laborglasinstrumente™ B0463061
Additional Details : Weight : 0.10000kg
Product Code. 10539030
  • 81.00€ / Each
Estimated Shipment: 21-06-2024
Description
  • According to ISO
  • Material: DURAN™ Borosilicate Glass
  • For use with liquid extractions
Specifications
Specifications
2.5 mm
250 mL
Duran Glass, Borosilicate Glass