Learn More
DWK Life Sciences DURAN™ Filter Disc Sintered Glass, Porosity 0, edge not fused
A prerequisite for successful work with glass filters is the selection of the right porosity. It is important to select a filter whose largest pore size is slightly below the grain size of the smallest particles still to be separated.
14.35€ - 80.35€
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
For Use With (Application) | Suitable for vacuum filtration with a maximum resulting pressure difference of 1 bar |
Product Code | Brand | Diameter (Metric)Top | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric)Top | Price | Quantity & Availability | |||||
10084860
|
DWK Life Sciences
251580001 |
80 mm |
29.80€
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
15202039
|
DWK Life Sciences
251540006 |
40 mm |
73.15€
Pack of 10 |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
11690009
|
DWK Life Sciences
251600009 |
100 mm |
71.90€
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
11639988
|
DWK Life Sciences
251535003 |
35 mm |
61.55€
Pack of 10 |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
11639998
|
DWK Life Sciences
251555005 |
55 mm |
14.35€
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
11669988
|
DWK Life Sciences
251545004 |
45 mm |
80.35€
Pack of 10 |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
11659978
|
DWK Life Sciences
251515001 |
15 mm |
49.40€
Pack of 10 |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
11600009
|
DWK Life Sciences
251565006 |
65 mm |
23.45€
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
15222019
|
DWK Life Sciences
251510003 |
10 mm |
47.95€
Pack of 10 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
15252029
|
DWK Life Sciences
251530005 |
30 mm |
56.15€
Pack of 10 |
Estimated Shipment: 22-08-2024
Log in to see stock. |
|||||
10589001
|
DWK Life Sciences
251550007 |
50 mm |
48.50€
Pack of 5 |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
10624182
|
DWK Life Sciences
251560008 |
60 mm |
15.60€
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
10203271
|
DWK Life Sciences
251570009 |
70 mm |
27.15€
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
10471263
|
DWK Life Sciences
251590002 |
90 mm |
33.70€
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
15202029
|
DWK Life Sciences
251525002 |
25 mm |
50.95€
Pack of 10 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
10519011
|
DWK Life Sciences
251620002 |
120 mm |
80.30€
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
Description
Glass powder with a certain particle size distribution results in a porous, solid body. These bodies are used as filter material in the form of plates and candles. Made of DURAN™ (borosilicate glass 3.3). They are ideally suited for separations with strong acids or alkalis, for example, and thus offer advantages over other materials such as plastic or paper. Available in 6 different porosity classes according to ISO 4796 as well as ASTM EI28-99. Suitable for vacuum filtration with a maximum resulting pressure difference of 1 bar.
- Edge non-melted and centered, surfaces untreated
- Successful work through the correct choice of porosity (The largest pore size should be slightly below the grain size of the smallest particles still to be separated).
- Available in 6 different porosity classes according to ISO 4796 as well as ASTM EI28-99.
- Glass articles for further processing
Specifications
Borosilicate Glass (3.3) | |
Suitable for vacuum filtration with a maximum resulting pressure difference of 1 bar |