Learn More
DWK Life Sciences DURAN™ Capillary-Single way Stopcock, ground and polished, without key
Ground and polished shut-off taps without plug for grease free operation.
1015.00€ - 1115.00€
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
Autoclavable | Yes |
No. per Pack | 50/Pk. |
For Use With (Application) | Coarse dosing of liquids; Installation in glass apparatus and devices |
Product Code | Brand | Closure Size | Plug Bore | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Size | Plug Bore | Price | Quantity & Availability | ||||
15894758
|
DWK Life Sciences
286440002 |
14.5 NS | 2.5 mm |
1115.00€
Pack of 50 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||||
15864758
|
DWK Life Sciences
286439207 |
12.5 NS | 2 mm |
1015.00€
Pack of 50 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||||
Description
Ground and polished DURAN™ single way stopcocks are shut-off taps which are combined with PTFE valve plugs to allow grease free operation.
- Ground shut-off taps without tap plug
- Fat-free working through the polished tap surface
- Taper 1:10
- Glass articles for further processing
Specifications
Borosilicate Glass (3.3) | |
Yes | |
50/Pk. | |
Coarse dosing of liquids; Installation in glass apparatus and devices |