Learn More
DWK Life Sciences DURAN™ Blank for separating funnel, conical for ISO 4800
For further processing as separating funnel with tap and standard ground joint.
65.05€ - 162.00€
Specifications
Material | Boro 3.3 |
---|---|
Autoclavable | Yes |
For Use With (Application) | Blank for further processing as dropping funnel; Phase separation |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15221629
|
DWK Life Sciences
210136302 |
2000 mL |
84.40€
Pack of 2 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
15281619
|
DWK Life Sciences
210132403 |
100 mL |
65.05€
Pack of 10 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
15291619
|
DWK Life Sciences
210133605 |
250 mL |
72.05€
Pack of 10 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
15211629
|
DWK Life Sciences
210135409 |
1000 mL |
162.00€
Pack of 10 |
In Stock
Log in to see stock. |
|||||
15201629
|
DWK Life Sciences
210134404 |
500 mL |
121.00€
Pack of 10 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
Description
DURAN™ separating funnels are used in chemistry to separate non-mixable liquids. DURAN™ separating funnels Raw bodies are designed for forming the appropriate standard ground joint sleeve according to DIN ISO 4800.
- Conical shape for ISO 4800
- Glass articles for further processing
Specifications
Boro 3.3 | |
Yes | |
Blank for further processing as dropping funnel; Phase separation |