Learn More
Abnova™ DPYD/CEN1p FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
1475.00€ - 2430.00€
Specifications
Content And Storage | Store at 4°C in the dark |
---|---|
Format | Tube |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Gene | dihydropyrimidine dehydrogenase |
Gene Alias | DHP,DHPDHASE,DPD,MGC132008,MGC70799 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16053355
|
Abnova™
FG0219.100UL |
100 μL |
1475.00€
100µL |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16063355
|
Abnova™
FG0219.200UL |
200 μL |
2430.00€
200µL |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
DHP,DHPDHASE,DPD,MGC132008,MGC70799 | |
1806 | |
Liquid |
Tube | |
dihydropyrimidine dehydrogenase | |
DPYD | |
DNA/RNA |