missing translation for 'onlineSavingsMsg'
Learn More

Domperidone, Thermo Scientific™

Peripheral dopamine receptor antagonist that does not cross the blood-brain barrier

82.20€ - 291.00€

Chemical Identifiers

CAS 57808-66-9
Molecular Formula C22H24ClN5O2
Molecular Weight (g/mol) 425.917
MDL Number MFCD00069256
InChI Key FGXWKSZFVQUSTL-UHFFFAOYSA-N
Synonym domperidone, motilium, nauzelin, domperidonum, domperidona, domperidon, domperidonum inn-latin, domperidona inn-spanish, motillium, motinorm
PubChem CID 3151
ChEBI CHEBI:31515
IUPAC Name 6-chloro-3-[1-[3-(2-oxo-3H-benzimidazol-1-yl)propyl]piperidin-4-yl]-1H-benzimidazol-2-one
SMILES C1CN(CCC1N2C3=C(C=C(C=C3)Cl)NC2=O)CCCN4C5=CC=CC=C5NC4=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15485859
SDS View Documents
Thermo Scientific Alfa Aesar
J63681.MC
100mg
82.20€
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15495859
SDS View Documents
Thermo Scientific Alfa Aesar
J63681.ME
500mg
291.00€
500mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Domperidone is used as a Peripheral dopamine receptor antagonist that does not cross the blood-brain barrier. The antidopaminergic effects of Domperidone have been correlated to antiemetic and gastroprokinetic action.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications

Chemical Identifiers

57808-66-9
425.917
FGXWKSZFVQUSTL-UHFFFAOYSA-N
3151
6-chloro-3-[1-[3-(2-oxo-3H-benzimidazol-1-yl)propyl]piperidin-4-yl]-1H-benzimidazol-2-one
C22H24ClN5O2
MFCD00069256
domperidone, motilium, nauzelin, domperidonum, domperidona, domperidon, domperidonum inn-latin, domperidona inn-spanish, motillium, motinorm
CHEBI:31515
C1CN(CCC1N2C3=C(C=C(C=C3)Cl)NC2=O)CCCN4C5=CC=CC=C5NC4=O

Specifications

57808-66-9
MFCD00069256
domperidone, motilium, nauzelin, domperidonum, domperidona, domperidon, domperidonum inn-latin, domperidona inn-spanish, motillium, motinorm
FGXWKSZFVQUSTL-UHFFFAOYSA-N
6-chloro-3-[1-[3-(2-oxo-3H-benzimidazol-1-yl)propyl]piperidin-4-yl]-1H-benzimidazol-2-one
3151
425.91
C22H24ClN5O2
14,3418
Slightly soluble in water.
C1CN(CCC1N2C3=C(C=C(C=C3)Cl)NC2=O)CCCN4C5=CC=CC=C5NC4=O
425.917
CHEBI:31515
Domperidone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H361
Suspected of damaging fertility or the unborn child.

P201-P202-P281-P308+P313-P501c

H361

missing translation for 'einecsNumber' : 260-968-7

missing translation for 'rtecsNumber' : DE2275900

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.