Learn More
MP Biomedicals™ Dome Clip/Latch for FastPrep-24™
Product Code. 13417278 Shop All MP Biomedicals Products
Change view
Click to view available options
For Use With (Equipment):
FastPrep-24 Instruments
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
13417278 FastPrep-24 Instruments Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13417278 Supplier MP Biomedicals™ Supplier No. 116002508
  • ONLINE EXCLUSIVE PRICE
    15.45€
    14.70€ / Pack of 1
    Save 0.75€
    4% Off
In Stock
Add to basket
Edge
Add to basket
Dome Clip/Latch for FastPrep-24™ is a replacement part for use with all models of FastPrep-24™ instruments.
  • Compatible with all models of FastPrep-24 instruments
Specifications
For Use With (Equipment) FastPrep-24 Instruments
Packaging Quantity 1 Ea.
Product Type Replacement Shaft Clips
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.