Learn More
Fisherbrand™ DNA Sequencing Systems: Combs for Discontinued Model FB-SEQ-2045
DNA Sequencing Systems Combs for discontinued model FB-SEQ-2045
Brand:  Fisherbrand™
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11933075
  • 53.85€ / Each
Estimated Shipment: 12-09-2024
Description
For use with discontinued model FB-SEQ-2045
Specifications
Specifications
Owl s4s aluminum backed sequencer system
50 cm
17.7 in.
45 cm
20
15μL
27 cm
19.6 in.
9 in.
23 cm
Sharktooth Comb
0.4 mm
10.6 in.
20 cm