Learn More
Thermo Scientific™ Viper™ Capillary Kits for LC Pumps
Order this single kit for all the easy, zero-dead-volume Thermo Scientific™ Viper™ Fingertight Fittings assembled with capillaries for your LC pumps.
Description
Avoid data-destroying dead volumes in the capillary connections of your HPLC or UHPLC pump connections by using Thermo Scientific™ Viper™ Capillary Kits for LC Pumps. This kit provides worry-free connections for Thermo Scientific™ Dionex™ UltiMate™ 3000 ISO, LPG, or DGP Pumps in UltiMate RSLC and SD systems. Even with conventional columns and moderate pressures, poor connections can cause retention time shifts and peak distortions. The kit contains ViperFingertight Fittings which seal at the tip of the tubing and eliminate dead volumes and make it easier to plumb your HPLC or UHPLC system.
Worry-Free ConnectionsYou won’t have to worry about capillary slippage or improper ferrule positioning anymore. Consistently perfect connections ensure robust chromatography with minimal interruption.
Pump-Specific Kits- UltiMate 3000 ISO, LPG, or DGP Pumps used in UltiMate 3000 SD Systems
- UltiMate 3000 HPG Pumps used in UltiMate 3000 SD Systems
- UltiMate 3000 ISO, LPG, or DGP Pumps used in UltiMate 3000 RS Systems
- UltiMate 3000 HPG Pumps forUltiMate 3000 RS Systems
Stainless-steel Viper capillaries (I.D. x L, one each):
- 0.18mm/0.007 in. x 450mm
- 0.13mm/0.005 in. x 350mm
- 0.13mm/0.005 in. x 250mm
Specifications
| Capillary Kit with Single System Connections |