Learn More
Trajan Scientific and Medical™ Diluter/Dispenser Syringes for V3 Pumps
Gives better performance and superior quality. Trajan Scientific and Medical Diluter/Dispenser Syringes for V3 Pumps has the knowledge and expertise that goes into manufacturing an SGE syringe.
98.40€ - 107.00€
Specifications
Description | 30mm stroke, 1/4-28 UNF thread connection, 4.8mm hole in plunger |
---|---|
For Use With (Equipment) | V3 Pump |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
15218513
|
Trajan Scientific and Medical™
008793 |
5 mL |
107.00€
Each |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
15268503
|
Trajan Scientific and Medical™
008593 |
2.5 mL |
104.00€
Each |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
15228503
|
Trajan Scientific and Medical™
008193 |
1 mL |
104.00€
Each |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
15278493
|
Trajan Scientific and Medical™
007693 |
500 μL |
98.40€
Each |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Diluter/Dispenser Syringe for V3 Pumps
Specifications
30mm stroke, 1/4-28 UNF thread connection, 4.8mm hole in plunger | |
V3 Pump |