Learn More
Trajan Scientific and Medical™ Diluter/Dispenser Syringes for Hamilton Microlab 500™
Gives better performance and superior quality. Trajan Scientific and Medical Diluter/Dispenser Syringes for Hamilton Microlab 500™ has the knowledge and expertise that goes into manufacturing an SGE syringe.
Brand:  Trajan Scientific and Medical™ 005232
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15238483
  • 77.50€ / Each
Estimated Shipment: 08-07-2024
to see stock.
Add to basket
Description
Description
Gives better performance and superior quality. Trajan Scientific and Medical Diluter/Dispenser Syringes for Hamilton Microlab 500™ has the knowledge and expertise that goes into manufacturing an SGE syringe.
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Diluter/Dispenser Syringe for Hamilton Microlab 500
Specifications
Specifications
60mm stroke, 6-32 UNC thread in plunger button
60 mm
100 μL
Plunger stop
Hamilton MicroLab 500™ Instrument Syringes
Luer Lock