missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl phosphite, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Description
Diethyl phosphite is precursor used for the synthesis of various organophosphorus compounds. used to prepare the nickel chloride-diethyl phosphite system, efficient catalyst for the cross-coupling reaction between various functionalized arylzinc halides and aryl bromides, triflates and activated chlorides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Spécification
Melting Point | -70°C |
Color | Colorless |
Density | 1.070 g/mL |
Boiling Point | 50°C to 51°C (2 mmHg) |
Flash Point | 74°C (165°F) |
Odor | Weak |
Quantity | 25 g |
Sensitivity | Moisture sensitive |
Solubility Information | Hydrolyzes with water. |
Formula Weight | 138.1 |
Afficher plus |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'