Learn More
Epredia™ Cytoblock™ Cell Block Preparation System
Facilitate quick, reliable processing of FNAs, tissue fragments, and cell buttons with the Cytoblock™ Cell Block Preparation System.
Brand: Epredia™ 7401150
204.79 EUR valid until 2024-03-29
Use promo code "21653" to get your promotional price.
Additional Details : Weight : 0.45000kg
Description
- Increases ability to process scanty or unseen material for cell block and reduces loss of critical material
- Suitable for immunohistochemical techniques
- Replaces time-consuming and costly agar and thrombin techniques
- Requires phosphate-free fixative on processor, such as Epredia™ fixatives Zinc Formal-Fixx™, Formal-Fixx, or Glyo-Fixx™ (formalin substitute) or simply bypass formalin fixation steps on processor
- Cytoblock system can also be run on a rapid biopsy program
Specifications
Cell Preparation System | |
Cell Block Preparation System |