Learn More
Kartell™ Plastilab™ All-Purpose Cylindrical Stirring Bars
Features rounded shape with smooth walls.
Brand:  Kartell™ 0075800
41.04 EUR valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10192661
  • 50.25€ / Pack of 10
Estimated Shipment: 27-05-2024
Description
Features rounded shape with smooth walls.
  • PTFE-coated magnet
  • Magnetic field causes stirring bar to spin very quickly
  • Non-stick, non-reactive and hydrophobic
  • Highly resistant to thermo-shock
  • Suitable for foodstuff
Specifications
Specifications
6.16 mm
10/Pk.
Cylinder, Sphere
Magnet PTFE Coated
29.88 mm