Learn More
Fisherbrand™ Stir Bar Box Set, Cylindrical
Product Code. 10226853 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Length (Metric):
195 mm
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10226853 195 mm 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10226853
  • ONLINE EXCLUSIVE PRICE
    63.60€
    36.44€ / 1 sets
    Save 27.16€
    42% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Provides general purpose stirring for a wide variety of applications. Fisherbrand™ Stir Bar Box Sets, Cylindrical are isostatically molded in pure, FDA-approved PTFE plastic.
  • Guaranteed inert and leak-free
  • Smooth, round profile
  • Individually polished to stay cleaner for longer
  • Each set contains two (2) each of the following sizes:
6 x 10mm
4.5 x 15mm
6 x 20mm
6 x 25mm
6 x 30mm
8 x 40mm
8 x 50mm
10 x 60mm
10 x 70mm
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color White
Description PTFE encapsulated Alnico V magnet core
Dimensions (L x W x H) 195 x 95mm
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Height (Metric) 30mm
Includes 18 Cylindrical Stirrer Bars
Width (Metric) 95mm
Length (Metric) 195 mm
Quantity 1/Ea.