Learn More
Multiroir™ Crystal Polystyrene Transmission Lab Box
Product Code. 11722944
Change view
Click to view available options
Capacity (Metric):
0.09 L
0.2 L
0.42 L
1.3 L
3.7 L
10.85 L
Unit Size:
Each
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11722944 0.2 L Pack of 10
11712944 0.09 L Pack of 10
11732944 0.42 L Pack of 10
11742944 1.3 L Each
11762944 10.85 L Each
11752944 3.7 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11722944 Supplier Multiroir™ Supplier No. 45102
Add to basket
Edge
Add to basket
Use these boxes for a variety of storage needs. Multiroir™ Crystal Polystyrene Transmission Lab Boxes are ideal for conservation, storage and presentation of products. Boxes are made of crystal polystyrene, a resistant material obtained by polymerization of styrene monomer CH2 = CH-Ph.
  • Transparent, stackable and highly resistant to UV
  • Integrated Label holder
  • Chemically inert plastic
  • Food contact quality
  • Funds and above have male-female reliefs that lock stacking and provide perfect storage stability
  • The tight tolerances of the fitting of the cover on the base ensure a good seal
Specifications
Capacity (Metric) 0.2 L
Color Transparent
Material Polystyrene
Shape Rectangular
For Use With (Application) Conservation, Storage and Presentation of Products
Product Type Transmission Lab Box
Quantity 10/Pk.
Dimensions (L x W x H) 60 L x 50 H mm
Length (Metric) 90 mm
Length (Metric) Interior 85 mm
Width (Metric) 60 mm
Height (Metric) 50 mm
Height (Metric) Interior 43 mm
Weight (Metric) 0.05 kg