Learn More
Multiroir™ Crystal Polystyrene Minimax Storage Boxes
Use Multiroir™ Crystal Polystyrene Minimax Storage Boxes for your storage and transport needs. The clear, glass-like polystyrene enables you to view your contents clearly from the outside. The boxes are delivered complete with a separator kit for additional organization of contents.
Marque: Multiroir™ 42051
6.49 EUR valid until 2024-05-31
Use promo code "21638" to get your promotional price.
Informations supplémentaires : Weight : 0.13500kg
Description
- Transparent as glass
- Delivered complete with separator kit
- Various capacities
Spécification
Transparent | |
Rectangular | |
1 Each | |
45 mm |
Polystyrene | |
Storage Box | |
178 mm |