Learn More
Saint Gobain Life Sciences Tygon™ LMT-55 Tubing with Three Stop Configuration
Formulated for resistance to flex fatigue and abrasion and is less permeable than rubber tubing. Tygon™ LMT-55 Tubing with Three Stop Configuration handles virtually all inorganic chemicals found in the lab.
42.52€ - 74.35€
Specifications
Min. Operating Temperature (Metric) | -50°C |
---|---|
MaxOperatingTempMetric | 74°C |
Product Code | Brand | Diameter (Metric) Inner | Thickness (Metric) | Color | Overall Length (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Thickness (Metric) | Color | Overall Length (Metric) | Price | Quantity & Availability | ||
12340160
|
Saint Gobain Life Sciences
070535-00-ND |
0.13 mm | 0.90 mm | Orange/Orange | 38.1 cm |
70.45€
Pack of 12 |
In Stock
Log in to see stock. |
||
11704648
|
Saint Gobain Life Sciences
070535-05-ND |
0.51 mm | 0.90 mm | Orange/Yellow | 38.1 cm |
70.45€
Pack of 12 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||
11714648
|
Saint Gobain Life Sciences
070535-07-ND |
0.64 mm | 0.90 mm | Orange/White | 38.1 cm |
70.45€
Pack of 12 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
15103216
|
Saint Gobain Life Sciences
070535-13-ND |
1.14 mm | 0.85 mm | Red/Red | 38.1 cm |
70.45€
Pack of 12 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||
11734648
|
Saint Gobain Life Sciences
070535-15-ND |
1.30 mm | 0.85 mm | Gray/Gray | 38.1 cm |
70.45€
Pack of 12 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
15183206
|
Saint Gobain Life Sciences
070535-18-ND |
1.65 mm | 0.85 mm | Blue/Blue | 38.1 cm |
74.35€
Pack of 12 |
N/A | ||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
11749744
|
Saint Gobain Life Sciences
070535-25-ND |
3.17 mm | 0.85 mm | Black/White | 38.1 cm |
74.35€
Pack of 12 |
N/A | |
Currently Unavailable: We have no information when this product will be available
View alternative products
|
10773816
|
Saint Gobain Life Sciences
070535-16-ND |
1.42 mm | 0.90 mm | Crystal Clear | 38.1 cm |
70.45€
Pack of 12 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|
13474815
|
Saint Gobain Life Sciences
070535-11-ND |
1.02 mm | 0.85 mm | Crystal Clear | 10 m |
70.45€
Pack of 12 |
In Stock
Log in to see stock. |
||
15123216
|
Saint Gobain Life Sciences
070535-23-ND |
2.54 mm | 0.85 mm | Crystal Clear | 10 m |
70.45€
Pack of 12 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||
15133216
|
Saint Gobain Life Sciences
070535-24-ND |
2.79 mm | 0.85 mm | Crystal Clear | 10 m |
70.45€
Pack of 12 |
N/A | ||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
15171773
|
Saint Gobain Life Sciences
070535-17-ND |
1.52 mm | 0.85 mm | Crystal Clear | 10 m |
70.45€
Pack of 12 |
N/A | |
Currently Unavailable: We have no information when this product will be available
View alternative products
|
10320881
|
Saint Gobain Life Sciences
SC0056T |
0.76 mm | 0.85 mm | Black/Black | 38.1 cm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||
Description
- Bio-based formulation delivers excellent performance
- Lot-to-lot consistency for reproducible results
- Non-toxic and non-contaminating
- Smooth, polished inner wall
- Slips easily over fittings and grips securely for simple lab set-ups
- Contains no BPA or phthalates
- Standard sizes available to hold full vacuum at room temperature
- General laboratory
- Food and beverage
- Biopharmaceutical
- Analytical instruments
- Peristaltic and vacuum pumps
- Ideal for condensers, incubators, desiccators, gas lines and drain line
Specifications
-50°C | |
74°C |