Learn More
Fisherbrand™ Crimping Heads for Pneumatic Hand-held Crimping Tool
For use with Pneumatic Hand-held Crimping Tool.
Brand: Fisherbrand™
726.75 EUR valid until 2024-12-31
Use promo code "23262" to get your promotional price.
Additional Details : Weight : 1.10000kg
Description
- Crimping Head
- Pneumatic
- Steel
Specifications
Flip Top/Flip Off Seals | |
20mm Crimping Head |