Learn More
Helios Italquartz Covers with Long Handle, Translucent
Covers with long handle constructed of translucent, durable, resistant, and chemically inert quartz glass.
Brand:  Helios Italquartz 64F00605
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15818074
  • 101.00€ / Each
Estimated Shipment: 22-07-2024
Description
  • Translucent
  • Include long handle
  • Chemically inert
  • Excellent electrical insulation
  • Excellent strength and durability
  • Exceptional thermal stability compared to glass
Specifications
Specifications
Translucent
Quartz
Covers with Long Handle
CR 13 Crucible