Learn More
Helios Italquartz Covers with Long Handle, Translucent
Covers with long handle constructed of translucent, durable, resistant, and chemically inert quartz glass.
Brand:  Helios Italquartz 64F00604
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15808074
  • 83.25€ / Each
Estimated Shipment: 18-07-2024
Description
  • Translucent
  • Include long handle
  • Chemically inert
  • Excellent electrical insulation
  • Excellent strength and durability
  • Exceptional thermal stability compared to glass
Specifications
Specifications
Translucent
Quartz
Covers with Long Handle
CR 10 Crucible