Learn More
Sartorius™ Cotton Gloves for Weights
Product Code. 13119250 Shop All Sartorius Products
Click to view available options
:
For use with calibration weights and standards
Product Code. 13119250
Brand: Sartorius™ YAW21
Prevent damage to the surface of calibration weights. Sartorius™ Cotton Gloves for Calibration Weights allow you to safely and properly handle finely machined calibration weights and maintain balance accuracy.
  • Cotton construction
  • Ensure accuracy of weight and mass standards
  • Eliminates fingerprints and resists contamination by dirt, moisture, and corrosion
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Calibration Weights and Standards
Color White
Material Cotton
Quantity 1 Pair/Pk.