Learn More
Corning™ Costar™ Spin-X™ Centrifuge Tube Filters, CA Membrane
Product Code. 10649351 Shop All Corning Products
Change view
Click to view available options
Pore Size:
0.22 μm
0.45 μm
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Pack of 96
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Sterility unitSize
10649351 0.45 μm Non-sterile Pack of 100
10310361 0.22 μm Sterile Pack of 96
10787632 0.45 μm Sterile Pack of 96
10104101 0.22 μm Non-sterile Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10649351 Supplier Corning™ Supplier No. 8163
  • ONLINE EXCLUSIVE PRICE
    209.00€
    195.21€ / Pack of 100
    Save 13.79€
    6% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Consisting of a membrane-containing filter unit within a polypropylene centrifuge tube. Corning™ Costar™ Spin-X™Centrifuge Tube Filters effectively filters small sample volumes by centrifugation for bacteria removal, particle removal, HPLC sample preparation, removal of cells from media and purification of DNA from agarose and polyacrylamide gels.
  • Spin-X centrifuge tube filters consist of a membrane-containing filter unit within a microcentrifuge tube.
  • Uses: Removing bacteria, cells, and particles from liquids; HPLC sample preparation; DNA removal from agarose or acrylamide gels. Maximum RCF is 16,000 x g.
TRUSTED_SUSTAINABILITY
Specifications
Material (Membrane) Cellulose Acetate
Pore Size 0.45 μm
Sterility Non-sterile
Type Spin-X
Volume (Metric) 0.5 mL