Learn More
Corning™ Reusable Phenolic Screw Caps With Rubber Liners
Click to view available options
Color:
Black
White
Description
These caps are fabricated of a resin specially formulated to resist the effects of temperature and steam in autoclaving. Caps have a glued-in, white rubber liner.
Specifications
Closure Material | Phenolic |
Closure Type | Rubber Liner |
Color | White |
Material | Phenolic Resin |
No. per Case | 144/Cs. |
No. per Pack | 144/Pk. |
Closure Color | Black |