Learn More
Corning™ Collagen I, Rat
Shop All Corning Products
Click to view available options
Quantity:
100 mg
1000 mg
Unit Size:
100mg
1g
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning offers a broad range of collagen types that are derived from multiple species. These collagens have been used to culture a variety of cell types to promote adhesion, growth, and/or differentiation.
Specifications
Specifications
| Concentration | 3 to 4 mg/mL |
| For Use With (Application) | Cell Attachment and Proliferation, Cell-specific Morphology and Function |
| Formulation | Liquid in 0.02N acetic acid |
| Quantity | 1000 mg |
| Source | Rat Tail Tendon |
| Product Type | Collagen, Rat Tail Type I |
| Content And Storage | 2°C to 8°C |
| Purity or Quality Grade | >90% by SDS-PAGE |