Learn More
Lauda™ Connecting Plug
For Use With: For Pt100 connection
Brand: Lauda™ EQS 014
Additional Details : Weight : 30.00000kg
Description
- 4-pin for Pt100 connection
- Connecting cables
Specifications
For Pt100 connection | |
Connecting Plug |