Learn More
Abeba™ Comfort 6813 Shoes
Stylish hand-sewn seams and PU sole with high slip resistance.
Brand:  Abeba™ 6813/37
Additional Details : Weight : 0.85000kg
 View more versions of this product
Product Code. 15662328
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Description
Description
  • CE EN ISO 20347:2012 OB A, E SRB
  • Smooth leather
  • Black
  • Slip resistance PU outsole sole
  • Replaceable Reflexor insole with nubs (article no. 3572)
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
  • Lining microfiber, respiration absorbing
  • Instep area with elastic band
  • Hand-sewn seam
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E SRB
Microfiber
37
Polyurethane
Black
Leather
Slip-resistant
6813