Learn More
Fisherbrand™ Combs for MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit, 2mm Thick
Product Code. 11813383
Change view
Click to view available options
No. of Wells:
1 Prep, 1 Marker
10
12
14
16
18
2 Prep, 2 Marker
20
28
30
35
4 Prep, 2 Marker
Volume (Metric) Sample:
20 μL
23 μL
25 μL
43 μL
54 μL
59 μL
81 μL
90 μL
243 μL
450 μL
990 μL
Unit Size:
Each
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Wells Volume (Metric) Sample Unit Size
11813383 14 59 μL Each
11863363 10 90 μL Each
11803373 10 59 μL Each
11833373 1 Prep, 1 Marker 990 μL Each
11873373 12 81 μL Each
11853383 16 54 μL Each
11893383 18 23 μL Each
11843393 20 43 μL Each
11873393 2 Prep, 2 Marker 450 μL Each
11813403 28 23 μL Each
11853403 30 25 μL Each
11893403 35 20 μL Each
11833413 4 Prep, 2 Marker 243 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 11813383
  • ONLINE EXCLUSIVE PRICE
    41.15€
    29.69€ / Pack of 1
    Save 11.46€
    27% Off
Estimated Shipment: 03-12-2026
Add to basket
Add to basket
For use in preparatory electrophoresis and high-throughput screening.
  • Recommended for high resolution gels and publication quality data
  • Maximise sample volume
  • Height adjustable to give user full control over well depth and sample-loading volume
  • Rigid back which prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
  • 10, 14, 16, 18, 28 and 30-sample combs are Multichannel pipettor compatible
Specifications
For Use With (Equipment) MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit
Type Comb
No. of Wells 14
Quantity 1 Each
Thickness (Metric) 2 mm
Volume (Metric) Sample 59 μL