Learn More
Fisherbrand™ Combs for Horizontal Tanks MultiSUB Midi Gel Electrophoresis Unit, 2mm Thick
For use in preparatory electrophoresis and high-throughput screening.
Brand: Fisherbrand™
Additional Details : Weight : 0.01000kg
Description
- Recommended to scale-up nucleic acid volumes for preparatory techniques
- Maximise sample volume
- Height adjustable to give user full control over well depth and sample-loading volume
- Rigid back which prevents heat-induced warping
- Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
- 10-sample and 20-sample combs are Multichannel pipettor compatible
Specifications
Sub-Gel Midi Horizontal Gel Electrophoresis Unit | |
2 Prep, 2 Marker | |
2 mm |
Comb | |
1 Each | |
315 μL x 8 |