Learn More
Lovibond™ COD Standard Solution
COD Standard Solution 100mg/L 30mL
Brand:  Lovibond™ 2420803
Additional Details : Weight : 0.30000kg
Product Code. 15382055
  • 34.20€ / Each
Estimated Shipment: 01-07-2024
Description
COD Standard Solution 100mg/L 30mL
Standard solutions are solutions with a defined concentration and are provided to check the operation methods and devices of the cuvette tests as well as the condition of the instrument.
Specifications
Specifications
100 mg/L
COD
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • STANDARD SOLUTION COD 100 MG/L
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid