Learn More
Tenak™ Clip-on Label Holder
Clip-on Label Holder for easy organizing of a freezer.
Brand: Tenak™ TE76100
Additional Details : Weight : 0.05000kg
Description
- Can be used for racks for both upright freezers and chest freezers
- Universal and can be used for most commonly sold racks of both stainless steel and aluminum with a material thickness of up to 1.5 mm
- Has a handel lifter which makes it easier to grab the rack handel (works with TENAK racks)
- Labels are not included
- Easy to install - Simply just clip it onto the rack and a barb secures it
Specifications
Racks for both upright freezers and chest freezers | |
1 Ea. | |
Clip-on Label Holder |