Learn More
Tenak™ Clip-on Label Holder
Clip-on Label Holder for easy organizing of a freezer.
Brand:  Tenak™ TE76120
Additional Details : Weight : 5.01000kg
 View more versions of this product
Product Code. 15211014
  • 1025.00€ / Pack of 100
Estimated Shipment: 04-10-2024
Description
  • Can be used for racks for both upright freezers and chest freezers
  • Universal and can be used for most commonly sold racks of both stainless steel and aluminum with a material thickness of up to 1.5 mm
  • Has a handel lifter which makes it easier to grab the rack handel (works with TENAK racks)
  • Labels are not included
  • Easy to install - Simply just clip it onto the rack and a barb secures it
Specifications
Specifications
Racks for both upright freezers and chest freezers
20/Pk.
Clip-on Label Holder